Diethyl iminodiacetate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Iminodiacetic Acid Diethyl Ester, Diethyl 2,2'-azanediyldiacetate

OR61269-100G
APOSOR61269-100GEA 42.2 EUR
APOSOR61269-100G
Diethyl iminodiacetate 98%
Diethyl iminodiacetate
Formel: C₈H₁₅NO₄
Molecular Weight: 189,21 g/mol
Siedepunkt: 208 °C
Dichte: 1,056 g/cm³ (25 °C)
Flammpunkt: 113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041925
CAS-Nummer: 6290-05-7
EINECS: 228-533-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR