N-Boc-L-alaninol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Boc-L-Alaninol, Boc-Alaninol, (S)-2-(BOC-amino)-1-propanol

OR61273-100G OR61273-25G OR61273-5G
APOSOR61273-100GEA 79.9 EUR
APOSOR61273-100G APOSOR61273-25G APOSOR61273-5G
N-Boc-L-alaninol 95%
N-Boc-L-alaninol
Formel: C₈H₁₇NO₃
Molecular Weight: 175,2 g/mol
Schmelzpunkt: 59…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00043121
CAS-Nummer: 79069-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR