5-Bromo-2-nitroaniline

Lieferant: Apollo Scientific
Achtung

OR61286-1G OR61286-25G OR61286-5G
APOSOR61286-1GEA 21.9 EUR
APOSOR61286-1G APOSOR61286-25G APOSOR61286-5G
5-Bromo-2-nitroaniline
5-Bromo-2-nitroaniline
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217.0200 g/mol
Schmelzpunkt: 151…153 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09056836
CAS-Nummer: 5228-61-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR