4-Amino-2,6-dichlorpyrimidin 98+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Dichloropyrimidin-4-ylamine
OR6129-25G
OR6129-5G
APOSOR6129-25GEA
26.1
EUR
APOSOR6129-25G
APOSOR6129-5G
4-Amino-2,6-dichlorpyrimidin 98+%
4-Amino-2,6-dichloropyrimidine
Formel:
C₄H₃Cl₂N₃ Molecular Weight: 163,99 g/mol Schmelzpunkt: 258…267 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00038015 CAS-Nummer: 10132-07-7 EINECS: 233-369-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...