3-Oxopentanenitrile 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR61297-25G
OR61297-5G
APOSOR61297-25GEA
524
EUR
APOSOR61297-25G
APOSOR61297-5G
3-Oxopentanenitrile 97%
3-Oxopentanenitrile
Formel:
C₅H₇NO Schmelzpunkt: 27…30 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07367087 CAS-Nummer: 33279-01-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...