3-Pyridazinamin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Pyridazin-3-amine

OR6130-1G OR6130-5G
APOSOR6130-1GEA 21.9 EUR
APOSOR6130-1G APOSOR6130-5G
3-Pyridazinamin 97%
3-Aminopyridazine
Formel: C4H5N3
Molecular Weight: 95,10 g/mol
Schmelzpunkt: 168…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01529869
CAS-Nummer: 5469-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR