3-Methoxy-4-nitrobenzenecarbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methoxy-4-nitrobenzonitrile
OR61302-1G
OR61302-250MG
OR61302-5G
APOSOR61302-1GEA
88.6
EUR
APOSOR61302-1G
APOSOR61302-250MG
APOSOR61302-5G
3-Methoxy-4-nitrobenzenecarbonitrile
3-Methoxy-4-nitrobenzenecarbonitrile
Formel:
C₈H₆N₂O₃ Molecular Weight: 178,15 g/mol Schmelzpunkt: 125…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11977368 CAS-Nummer: 177476-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...