4-Brom-3-nitrobenzonitril 98%

Lieferant: Apollo Scientific
Achtung

OR61310-10G OR61310-1G OR61310-5G
APOSOR61310-10GEA 62.5 EUR
APOSOR61310-10G APOSOR61310-1G APOSOR61310-5G
4-Brom-3-nitrobenzonitril 98%
4-Brom-3-nitrobenzonitril
Formel: C₇H₃BrN₂O₂
Molecular Weight: 227,02 g/mol
Schmelzpunkt: 116…121 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00982006
CAS-Nummer: 89642-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR