3-Amino-1-N-Cbz-piperidine 97%
Lieferant: Apollo Scientific
Synonyme:
3-Aminopiperidine, N1-CBZ protected
OR6134-10G
OR6134-25G
OR6134-5G
APOSOR6134-10GEA
139
EUR
APOSOR6134-10G
APOSOR6134-25G
APOSOR6134-5G
3-Amino-1-N-Cbz-piperidine 97%
3-Amino-1-N-Cbz-piperidine
Formel:
C13H18N2O2 Lagertemperatur: Raumtemperatur |
CAS-Nummer:
711002-74-3 UN: 1760 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...