3-Chloro-5-cyano-2-pyridine carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR61372-1G OR61372-500MG
APOSOR61372-1GEA 865 EUR
APOSOR61372-1G APOSOR61372-500MG
3-Chloro-5-cyano-2-pyridine carboxylic acid
3-Chloro-5-cyano-2-pyridine carboxylic acid
Formel: C₇H₃ClN₂O₂
Molecular Weight: 182,57 g/mol
Schmelzpunkt: 144 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD14584817
CAS-Nummer: 1200497-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR