2-Brom-4-methoxypyridin 98%

Lieferant: Apollo Scientific
Achtung

OR61413-1G OR61413-25G OR61413-5G
APOSOR61413-1GEA 30.6 EUR
APOSOR61413-1G APOSOR61413-25G APOSOR61413-5G
2-Brom-4-methoxypyridin 98%
2-Bromo-4-methoxypyridine
Formel: C₆H₆BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD06254613
CAS-Nummer: 89488-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR