2,7-Dibromfluoren

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,7-Dibromo-9H-fluorene

OR6144-100G OR6144-25G OR6144-250G OR6144-500G
APOSOR6144-100GEA 56.7 EUR
APOSOR6144-100G APOSOR6144-25G APOSOR6144-250G APOSOR6144-500G
2,7-Dibromfluoren
2,7-Dibromfluoren
Formel: C₁₃H₈Br₂
Molecular Weight: 324,01 g/mol
Schmelzpunkt: 164…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00019048
CAS-Nummer: 16433-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR