3,5-Dichloropicolinaldehyde 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dichloropyridine-2-carboxaldehyde

OR61442-1G OR61442-5G OR61442-250MG
APOSOR61442-1GEA 68.3 EUR
APOSOR61442-1G APOSOR61442-5G APOSOR61442-250MG
3,5-Dichloropicolinaldehyde 96%
3,5-Dichloropicolinaldehyde
Formel: C6H3Cl2NO
Molecular Weight: 176.0001 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09839149
CAS-Nummer: 471909-65-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR