3-Brom-4-iodanisol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-1-iodo-4-methoxybenzene
OR61448-1G
OR61448-5G
OR61448-25G
APOSOR61448-1GEA
24.8
EUR
APOSOR61448-1G
APOSOR61448-5G
APOSOR61448-25G
3-Brom-4-iodanisol 97%
3-Brom-4-iodanisol
Formel:
C₇H₆BrIO Molecular Weight: 312,9304 g/mol Siedepunkt: 90 °C/2mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD12547848 CAS-Nummer: 466639-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...