N-(2,4-Dichlorphenyl)-2-chloracetamid

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-N-(2,4-dichlorophenyl)acetamide, N-(Chloroacetyl)-2,4-dichloroaniline

OR6151-1G OR6151-5G
APOSOR6151-1GEA 178 EUR
APOSOR6151-1G APOSOR6151-5G
N-(2,4-Dichlorphenyl)-2-chloracetamid
N-(2,4-Dichlorphenyl)-2-chloracetamid
Formel: C₈H₆Cl₃NO
Molecular Weight: 238.498 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00018888
CAS-Nummer: 6974-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR