1-Chloroisoquinolin-4-ol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Chloro-4-hydroxyisoquinoline
OR61534-1G
OR61534-2.5G
OR61534-250MG
APOSOR61534-1GEA
183
EUR
APOSOR61534-1G
APOSOR61534-2.5G
APOSOR61534-250MG
1-Chloroisoquinolin-4-ol
1-Chloroisoquinolin-4-ol
Formel:
C₉H₆ClNO Molecular Weight: 179,61 g/mol Schmelzpunkt: 184…188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425941 CAS-Nummer: 3336-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...