2,3-Dihydrobenzo[1,4]dioxine-2-carbonitrile 95%

Lieferant: Apollo Scientific
Achtung

OR6154-1G OR6154-5G
APOSOR6154-1GEA 140 EUR
APOSOR6154-1G APOSOR6154-5G
2,3-Dihydrobenzo[1,4]dioxine-2-carbonitrile 95%
2,3-Dihydrobenzo[1,4]dioxine-2-carbonitrile
Formel: C₉H₇NO₂
Molecular Weight: 161.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00068066
CAS-Nummer: 1008-92-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR