3,4-Dibromiodbenzol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2-Dibromo-4-iodobenzene
OR61541-1G
OR61541-25G
OR61541-5G
APOSOR61541-1GEA
55.1
EUR
APOSOR61541-1G
APOSOR61541-25G
APOSOR61541-5G
3,4-Dibromiodbenzol 98%
3,4-Dibromiodbenzol
Formel:
C₆H₃Br₂I Lagertemperatur: Raumtemperatur |
MDL:
MFCD09753707 CAS-Nummer: 909072-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...