3-Bromo-N-methyl-2-nitroaniline
Lieferant: Apollo Scientific
|
Achtung
|
OR61607-1G
OR61607-250MG
APOSOR61607-1GEA
104
EUR
APOSOR61607-1G
APOSOR61607-250MG
3-Bromo-N-methyl-2-nitroaniline
3-Bromo-N-methyl-2-nitroaniline
Formel:
C₇H₇BrN₂O₂ Molecular Weight: 231.06 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12024389 CAS-Nummer: 1150617-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...