3-Bromo-1-N-methylbenzene-1,2-diamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-N1-methylbenzene-1,2-diamine

OR61609-1G OR61609-250MG
APOSOR61609-1GEA 374 EUR
APOSOR61609-1G APOSOR61609-250MG
3-Bromo-1-N-methylbenzene-1,2-diamine
3-Bromo-1-N-methylbenzene-1,2-diamine
Formel: C₇H₉BrN₂
Molecular Weight: 201,07 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12024390
CAS-Nummer: 1150617-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR