3-Acetyl-2-chlorpyridin 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-acetyl-2-chloropyridine, 1-(2-Chloro-3-pyridinyl)-1-ethanone, 1-(2-Chloropyridin-3-yl)ethanone

OR61610-1G OR61610-5G OR61610-10G
APOSOR61610-1GEA 21.9 EUR
APOSOR61610-1G APOSOR61610-5G APOSOR61610-10G
3-Acetyl-2-chlorpyridin 97%
3-Acetyl-2-chlorpyridin
Formel: C₇H₆ClNO
Molecular Weight: 155.5816 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840751
CAS-Nummer: 55676-21-6
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR