2-Formyl-4-picolin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Methylpyridine-2-carboxaldehyde, 4-Methylpicolinaldehyde
OR6171-1G
OR6171-5G
APOSOR6171-1GEA
0
EUR
APOSOR6171-1G
APOSOR6171-5G
2-Formyl-4-picolin 98%
2-Formyl-4-picolin
Formel:
C₇H₇NO Molecular Weight: 121,1366 g/mol Siedepunkt: 82…85 °C/11mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD07367933 CAS-Nummer: 53547-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...