5-Brom-2-formylpyridin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromopyridine-2-carbaldehyde, 5-Bromopicolinaldehyde, 5-Bromopyridine-2-carboxaldehyde

OR6176-25G OR6176-5G OR6176-100G
APOSOR6176-25GEA 88.6 EUR
APOSOR6176-25G APOSOR6176-5G APOSOR6176-100G
5-Brom-2-formylpyridin 97%
5-Brom-2-formylpyridin
Formel: C₆H₄BrNO
Molecular Weight: 186,01 g/mol
Schmelzpunkt: 91…96 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04112535
CAS-Nummer: 31181-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR