3-Brom-4-formylpyridin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-4-pyridinecarboxaldehyde, 3-Bromoisonicotinaldehyde, 3-Bromopyridine-4-carboxaldehyde

OR6177-1G OR6177-25G
APOSOR6177-1GEA 0 EUR
APOSOR6177-1G APOSOR6177-25G
3-Brom-4-formylpyridin 95%
3-Brom-4-formylpyridin
Formel: C₆H₄BrNO
Molecular Weight: 186,01 g/mol
Schmelzpunkt: 81…82 °C
Lagertemperatur: Kühlschrank
MDL: MFCD05864506
CAS-Nummer: 70201-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR