Benzyl-2-formylpyrrolidine-1-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Pyrrolidine-2-carboxaldehyde, N-CBZ protected

OR6182-500MG
APOSOR6182-500MGEA 328 EUR
APOSOR6182-500MG
Benzyl-2-formylpyrrolidine-1-carboxylate 95%
Benzyl-2-formylpyrrolidine-1-carboxylate
Formel: C₁₃H₁₅NO₃
Molecular Weight: 233,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115610
CAS-Nummer: 105706-84-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR