6-Chinolincarbaldehyd 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Formylquinoline

OR6184-1G OR6184-250MG
APOSOR6184-1GEA 21.9 EUR
APOSOR6184-1G APOSOR6184-250MG
6-Chinolincarbaldehyd 97%
6-Quinolinecarboxaldehyde
Formel: C₁₀H₇NO
Molecular Weight: 157,1700 g/mol
Schmelzpunkt: 75…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00805836
CAS-Nummer: 4113-04-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR