(2-Methyl-1,3-thiazol-4-yl)methylamine 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(Aminomethyl)-2-methyl-1,3-thiazole, 2-Methyl-1,3-thiazole-4-methylamine

OR6203-1G OR6203-250MG
APOSOR6203-1GEA 206 EUR
APOSOR6203-1G APOSOR6203-250MG
(2-Methyl-1,3-thiazol-4-yl)methylamine 97%
(2-Methyl-1,3-thiazol-4-yl)methylamine
Formel: C₅H₈N₂S
Molecular Weight: 128,2 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06212804
CAS-Nummer: 103694-26-4
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR