Methyl-5-chloro-2,6-dihydroxy-4-pyrimidinecarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 5-chloro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate

OR62051-1G OR62051-500MG
APOSOR62051-1GEA 459 EUR
APOSOR62051-1G APOSOR62051-500MG
Methyl-5-chloro-2,6-dihydroxy-4-pyrimidinecarboxylate
Methyl-5-chloro-2,6-dihydroxy-4-pyrimidinecarboxylate
Formel: C₆H₅ClN₂O₄
Molecular Weight: 204.57 g/mol
Schmelzpunkt: 262…263 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12025873
CAS-Nummer: 91447-90-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR