4-Chloro-5-iodo-6-methyl-2-pyrimidinamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-chloro-5-iodo-6-methylpyrimidine

OR62053-1G OR62053-500MG
APOSOR62053-1GEA 157 EUR
APOSOR62053-1G APOSOR62053-500MG
4-Chloro-5-iodo-6-methyl-2-pyrimidinamine
4-Chloro-5-iodo-6-methyl-2-pyrimidinamine
Formel: C₅H₅ClIN₃
Molecular Weight: 269,47 g/mol
Schmelzpunkt: 197…199 °C
Lagertemperatur: Kühlschrank
MDL: MFCD12024467
CAS-Nummer: 897030-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR