2-Brom-3-nitroanisol 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-methoxy-3-nitrobenzene

OR6218-5G OR6218-25G OR6218-1G
APOSOR6218-5GEA 29 EUR
APOSOR6218-5G APOSOR6218-25G APOSOR6218-1G
2-Brom-3-nitroanisol 99%
2-Brom-3-nitroanisol
Formel: C₇H₆BrNO₃
Molecular Weight: 232.0300 g/mol
Schmelzpunkt: 93…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07779272
CAS-Nummer: 67853-37-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR