4-(Aminomethyl)phenylboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(Aminomethyl)phenyl)boronic acid

OR6222-1G OR6222-250MG
APOSOR6222-1GEA 458 EUR
APOSOR6222-1G APOSOR6222-250MG
4-(Aminomethyl)phenylboronic acid
4-(Aminomethyl)phenylboronic acid
Formel: C₇H₁₀BNO₂
Schmelzpunkt: 163…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06213214
CAS-Nummer: 51239-46-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR