2-Methyl-5-nitroanisol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methoxy-2-methyl-5-nitrobenzene, Methyl 5-nitro-o-tolyl ether, 2-Methoxy-4-nitrotoluene
OR6224-500G
APOSOR6224-500GEA
201
EUR
APOSOR6224-500G
2-Methyl-5-nitroanisol 98%
2-Methyl-5-nitroanisol
Formel:
C₈H₉NO₃ Molecular Weight: 167,16 g/mol Schmelzpunkt: 69…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043912 CAS-Nummer: 13120-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...