4-Phenyl-2-pyridine carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR6247-1G OR6247-50MG OR6247-500MG OR6247-5G
APOSOR6247-1GEA 380 EUR
APOSOR6247-1G APOSOR6247-50MG APOSOR6247-500MG APOSOR6247-5G
4-Phenyl-2-pyridine carboxylic acid
4-Phenyl-2-pyridine carboxylic acid
Formel: C₁₂H₉NO₂
Molecular Weight: 199.21 g/mol
Schmelzpunkt: 154…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114164
CAS-Nummer: 52565-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR