4-(N-Methylpiperazino)-2-pyridine carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR6250-1G OR6250-250MG
APOSOR6250-1GEA 314 EUR
APOSOR6250-1G APOSOR6250-250MG
4-(N-Methylpiperazino)-2-pyridine carboxylic acid 97%
4-(N-Methylpiperazino)-2-pyridine carboxylic acid
Formel: C₁₁H₁₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08056300
CAS-Nummer: 914637-18-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR