4-Imidazolo-2-pyridine carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR6251-250MG
OR6251-5G
OR6251-100MG
OR6251-500MG
APOSOR6251-250MGEA
223
EUR
APOSOR6251-250MG
APOSOR6251-5G
APOSOR6251-100MG
APOSOR6251-500MG
4-Imidazolo-2-pyridine carboxylic acid 97%
4-Imidazolo-2-pyridine carboxylic acid
Formel:
C₉H₇N₃O₂ Molecular Weight: 189.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08056301 CAS-Nummer: 914637-20-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...