2-(1H-Imidazol-1-yl)isonicotinic acid 97%

Lieferant: Apollo Scientific
Achtung

OR6259-1G OR6259-250MG OR6259-500MG OR6259-2.5G
APOSOR6259-1GEA 366 EUR
APOSOR6259-1G APOSOR6259-250MG APOSOR6259-500MG APOSOR6259-2.5G
2-(1H-Imidazol-1-yl)isonicotinic acid 97%
2-(1H-Imidazol-1-yl)isonicotinic acid
Formel: C₉H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08235205
CAS-Nummer: 914637-28-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR