N-Hexanoyl-D-erythro-sphingosine

Lieferant: Apollo Scientific
Achtung

Synonyme: C6 ceramide, Caproyl ceramide, N-((2S,3R,E)-1,3-Dihydroxyoctadec-4-en-2-yl)hexanamide

OR6260T-25MG OR6260T-5MG
APOSOR6260T-25MGEA 275 EUR
APOSOR6260T-25MG APOSOR6260T-5MG
N-Hexanoyl-D-erythro-sphingosine
N-Hexanoyl-D-erythro-sphingosine

A biologically active, cell permeable, nonphysiologic ceramide analog. It stimulates protein phosphatase 2A at concentrations as low as 10nM & activiates MAP kinase.

Formel: C₂₄H₄₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00870174
CAS-Nummer: 124753-97-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR