5-Brom-6-chlornicotinsäure 97%

Lieferant: Apollo Scientific
Achtung

OR6264-1G OR6264-25G OR6264-5G
APOSOR6264-1GEA 21.9 EUR
APOSOR6264-1G APOSOR6264-25G APOSOR6264-5G
5-Brom-6-chlornicotinsäure 97%
5-Bromo-6-chloronicotinic acid
Formel: C₆H₃BrClNO₂
Molecular Weight: 236.45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01927098
CAS-Nummer: 29241-62-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR