2-(2-Furyl)-1,3-thiazole-4-carboxylic acid
Lieferant: Apollo Scientific
OR6275-1G
OR6275-250MG
OR6275-500MG
APOSOR6275-1GEA
432
EUR
APOSOR6275-1G
APOSOR6275-250MG
APOSOR6275-500MG
2-(2-Furyl)-1,3-thiazole-4-carboxylic acid
2-(2-Furyl)-1,3-thiazole-4-carboxylic acid
Formel:
C₈H₅NO₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD08235208 CAS-Nummer: 115299-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...