5-(2-Furyl)-1,3-oxazole-4-carboxylic acid 95%

Lieferant: Apollo Scientific

OR6280-1G OR6280-250MG OR6280-5G OR6280-50MG OR6280-500MG OR6280-2.5G
APOSOR6280-1GEA 209 EUR
APOSOR6280-1G APOSOR6280-250MG APOSOR6280-5G APOSOR6280-50MG APOSOR6280-500MG APOSOR6280-2.5G
5-(2-Furyl)-1,3-oxazole-4-carboxylic acid 95%
5-(2-Furyl)-1,3-oxazole-4-carboxylic acid
Formel: C₈H₅NO₄
Molecular Weight: 179,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07376160
CAS-Nummer: 143659-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR