5-Thien-2-yl-1,3-oxazole-4-carboxylic acid 95%

Lieferant: Apollo Scientific

OR6282-1G OR6282-250MG OR6282-5G OR6282-50MG OR6282-500MG
APOSOR6282-1GEA 209 EUR
APOSOR6282-1G APOSOR6282-250MG APOSOR6282-5G APOSOR6282-50MG APOSOR6282-500MG
5-Thien-2-yl-1,3-oxazole-4-carboxylic acid 95%
5-Thien-2-yl-1,3-oxazole-4-carboxylic acid
Formel: C₈H₅NO₃S
Molecular Weight: 195.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07376148
CAS-Nummer: 143659-15-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR