6-(4-Morpholinyl)-2-pyrazinecarboxylic acid 95+%
Lieferant: Apollo Scientific
Synonyme:
6-(4-Morpholinyl)pyrazine-2-carboxylic acid
OR6290-250MG
OR6290-1G
OR6290-5G
APOSOR6290-250MGEA
85.7
EUR
APOSOR6290-250MG
APOSOR6290-1G
APOSOR6290-5G
6-(4-Morpholinyl)-2-pyrazinecarboxylic acid 95+%
6-(4-Morpholinyl)-2-pyrazinecarboxylic acid
Formel:
C₉H₁₁N₃O₃ Molecular Weight: 209,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00233274 CAS-Nummer: 40262-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...