6-Chloropyridazine-3-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Carboxy-6-chloropyridazine, 3-Carboxy-6-chloro-1,2-diazine

OR6298-5G OR6298-1G OR6298-500MG OR6298-10G
APOSOR6298-5GEA 65.4 EUR
APOSOR6298-5G APOSOR6298-1G APOSOR6298-500MG APOSOR6298-10G
6-Chloropyridazine-3-carboxylic acid 97%
6-Chloropyridazine-3-carboxylic acid
Formel: C₅H₃ClN₂O₂
Molecular Weight: 158.54 g/mol
Schmelzpunkt: 145…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00160464
CAS-Nummer: 5096-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR