6-Methyl-3-oxo-2,3-dihydropyridazine-4-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Dihydro-6-methyl-3-oxopyridazine-4-carboxylic acid
OR6302-1G
OR6302-250MG
OR6302-5G
APOSOR6302-1GEA
108
EUR
APOSOR6302-1G
APOSOR6302-250MG
APOSOR6302-5G
6-Methyl-3-oxo-2,3-dihydropyridazine-4-carboxylic acid 97%
6-Methyl-3-oxo-2,3-dihydropyridazine-4-carboxylic acid
Formel:
C₆H₆N₂O₃ Molecular Weight: 154.13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08235213 CAS-Nummer: 74557-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...