Methyl-4-ethinylbenzoat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-ethynylbenzoate

OR63024-1G OR63024-25G OR63024-5G
APOSOR63024-1GEA 21.9 EUR
APOSOR63024-1G APOSOR63024-25G APOSOR63024-5G
Methyl-4-ethinylbenzoat 98%
Methyl-4-ethinylbenzoat
Formel: C₁₀H₈O₂
Molecular Weight: 160.17 g/mol
Schmelzpunkt: 92…95 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00168820
CAS-Nummer: 3034-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR