4-Chlor-3-iodbenzoesäure 97%
Lieferant: Apollo Scientific
OR63026-100G
APOSOR63026-100GEA
126
EUR
APOSOR63026-100G
4-Chlor-3-iodbenzoesäure 97%
4-Chloro-3-iodobenzoic acid
Formel:
C₇H₄ClIO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079732 CAS-Nummer: 42860-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...