3-Amino-5-brombenzonitril 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-3-bromobenzonitrile
OR63032-1G
OR63032-5G
OR63032-10G
APOSOR63032-1GEA
26.1
EUR
APOSOR63032-1G
APOSOR63032-5G
APOSOR63032-10G
3-Amino-5-brombenzonitril 99%
3-Amino-5-brombenzonitril
Formel:
C₇H₅BrN₂ Molecular Weight: 197,0300 g/mol Schmelzpunkt: 124…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109874 CAS-Nummer: 49674-16-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...