1H-Benzimidazol-4-carbonsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1H-Benzimidazole-7-carboxylic acid

OR6307-1G OR6307-250MG
APOSOR6307-1GEA 58 EUR
APOSOR6307-1G APOSOR6307-250MG
1H-Benzimidazol-4-carbonsäure 97%
1H-Benzimidazol-4-carbonsäure
Formel: C₈H₆N₂O₂
Molecular Weight: 162.14 g/mol
Schmelzpunkt: 326…330 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01823426
CAS-Nummer: 46006-36-4
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR