3-Chinolincarbonsäure 97%

Lieferant: Apollo Scientific
Achtung

OR6309-100G OR6309-25G
APOSOR6309-100GEA 429 EUR
APOSOR6309-100G APOSOR6309-25G
3-Chinolincarbonsäure 97%
3-Chinolincarbonsäure
Formel: C₁₀H₇NO₂
Molecular Weight: 173,17 g/mol
Schmelzpunkt: 277…283 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006770
CAS-Nummer: 6480-68-8
EINECS: 229-337-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR