3-Thiomorpholinecarboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR6324-1G OR6324-250MG
APOSOR6324-1GEA 197 EUR
APOSOR6324-1G APOSOR6324-250MG
3-Thiomorpholinecarboxylic acid 97%
3-Thiomorpholinecarboxylic acid
Formel: C₅H₉NO₂S
Molecular Weight: 147,2 g/mol
Schmelzpunkt: 270…278 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02181162
CAS-Nummer: 20960-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR